Assay Details
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057193.2 | NP_476541.2 | 5 - 6 | 751 | 127 | |
XM_006242864.2 | XP_006242926.1 | 5 - 6 | 929 | 127 | ||
GenBank mRNA | AJ305049.1 | - | 5 - 6 | 751 | 127 | |
FQ223464.1 | - | 5 - 6 | 772 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
