Assay Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013181.1 | NP_001013199.1 | 1 - 2 | 71 | 153 | |
GenBank mRNA | AY781102.1 | - | 1 - 2 | 71 | 153 | |
AY781103.1 | - | 1 - 2 | 71 | 153 | ||
AY781104.1 | - | 1 - 2 | 62 | 153 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY781102.1, AY781103.1, AY781104.1, NM_001013181.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
