Assay Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013181.1 | NP_001013199.1 | 3 - 4 | 1524 | 136 | |
GenBank mRNA | AB568265.1 | - | 2 - 3 | 322 | 136 | |
AY781102.1 | - | 3 - 4 | 1524 | 136 | ||
AY781103.1 | - | 3 - 4 | 1524 | 136 | ||
AY781104.1 | - | 3 - 4 | 1515 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
