Assay Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019179.1 | NP_062052.1 | 6 - 7 | 784 | 100 | |
XM_008767420.1 | XP_008765642.1 | 6 - 7 | 1273 | 100 | ||
XM_008767421.1 | XP_008765643.1 | 7 - 8 | 829 | 100 | ||
GenBank mRNA | BC126093.1 | - | 6 - 7 | 798 | 100 | |
L12138.1 | - | 6 - 7 | 784 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
