Assay Details
Gene Symbol: Epb41l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053927.1 | NP_446379.1 | 11 - 12 | 1425 | 118 | |
XM_006245651.2 | XP_006245713.1 | 11 - 12 | 3786 | 118 | ||
XM_008767414.1 | XP_008765636.1 | 11 - 12 | 3798 | 118 | ||
XM_008767415.1 | XP_008765637.1 | 11 - 12 | 3785 | 118 | ||
XM_008767416.1 | XP_008765638.1 | 11 - 12 | 3785 | 118 | ||
XM_008767417.1 | XP_008765639.1 | 11 - 12 | 3785 | 118 | ||
GenBank mRNA | AB032827.1 | - | 11 - 12 | 1425 | 118 | |
DQ462202.1 | - | 10 - 11 | 1345 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
