Assay Details
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245658.2 | XP_006245720.1 | 4 - 5 | 1475 | 85 | |
XM_006245661.2 | XP_006245723.1 | 3 - 4 | 1413 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
