Assay Details
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053395.1 | NP_445847.1 | 1 - 2 | 172 | 112 | |
GenBank mRNA | AF364071.1 | - | 1 - 2 | 172 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF364071.1, NM_053395.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
