Assay Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 5 - 6 | 468 | 87 | |
GenBank mRNA | FM118664.1 | - | 5 - 6 | 452 | 87 | |
FQ215476.1 | - | 5 - 6 | 463 | 87 | ||
FQ216044.1 | - | 5 - 6 | 467 | 87 | ||
FQ216100.1 | - | 5 - 6 | 444 | 87 | ||
FQ216185.1 | - | 5 - 6 | 464 | 87 | ||
FQ224925.1 | - | 5 - 6 | 465 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
