Assay Details
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017217.1 | NP_058913.1 | 7 - 8 | 1310 | 74 | |
XM_006257059.2 | XP_006257121.1 | 8 - 9 | 1640 | 74 | ||
GenBank mRNA | AB000113.1 | - | 7 - 8 | 1310 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
