Assay Details
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007667.2 | NP_001007668.1 | 1 - 2 | 356 | 153 | |
XM_008773228.1 | XP_008771450.1 | 1 - 2 | 286 | 153 | ||
XM_008773229.1 | XP_008771451.1 | 1 - 2 | 281 | 153 | ||
XM_008773230.1 | XP_008771452.1 | 1 - 2 | 281 | 153 | ||
GenBank mRNA | AY325208.1 | - | 221 | 153 | ||
BC060588.1 | - | 1 - 2 | 358 | 153 | ||
FQ222039.1 | - | 1 - 2 | 238 | 153 | ||
FQ229984.1 | - | 1 - 2 | 240 | 153 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008773228.1, XM_008773229.1, XM_008773230.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
