Assay Details
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007667.2 | NP_001007668.1 | 3 - 4 | 492 | 95 | |
GenBank mRNA | AY325208.1 | - | 357 | 95 | ||
BC060588.1 | - | 3 - 4 | 494 | 95 | ||
FQ222039.1 | - | 3 - 4 | 374 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
