Assay Details
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007667.2 | NP_001007668.1 | 4 - 5 | 596 | 106 | |
XM_008773228.1 | XP_008771450.1 | 4 - 5 | 1596 | 106 | ||
XM_008773229.1 | XP_008771451.1 | 4 - 5 | 992 | 106 | ||
XM_008773230.1 | XP_008771452.1 | 5 - 6 | 631 | 106 | ||
GenBank mRNA | AY325208.1 | - | 461 | 106 | ||
BC060588.1 | - | 4 - 5 | 598 | 6921572 | 106 | |
FQ222039.1 | - | 4 - 5 | 478 | 106 | ||
FQ229984.1 | - | 4 - 5 | 480 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
