Assay Details
Gene Symbol: Prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191888.1 | NP_001178817.1 | 1 - 2 | 74 | 67 | |
XM_006256963.2 | XP_006257025.1 | 1 - 2 | 141 | 67 | ||
XM_006256965.2 | XP_006257027.1 | 1 - 2 | 141 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001191888.1, XM_006256963.2, XM_006256965.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
