Assay Details
Gene Symbol: Prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191888.1 | NP_001178817.1 | 3 - 4 | 199 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001191888.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
