Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 23 - 24 | 2890 | 72 | |
XM_006257114.1 | XP_006257176.1 | 23 - 24 | 2906 | 72 | ||
XM_343797.7 | XP_343798.2 | 22 - 23 | 3005 | 72 | ||
GenBank mRNA | AF155826.1 | - | 1 - 2 | 127 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
