Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 5 - 6 | 795 | 111 | |
XM_006257114.1 | XP_006257176.1 | 5 - 6 | 811 | 111 | ||
XM_343797.7 | XP_343798.2 | 4 - 5 | 910 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
