Assay Details
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173151.1 | NP_775174.1 | 4 - 5 | 493 | 89 | |
XM_006257016.2 | XP_006257078.1 | 4 - 5 | 454 | 89 | ||
XM_006257017.2 | XP_006257079.1 | 4 - 5 | 861 | 89 | ||
XM_008773254.1 | XP_008771476.1 | 4 - 5 | 574 | 89 | ||
GenBank mRNA | AF190256.1 | - | 4 - 5 | 493 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
