Assay Details
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023957.1 | NP_076447.1 | 10 - 11 | 1887 | 107 | |
GenBank mRNA | AJ250425.1 | - | 10 - 11 | 1887 | 107 |
Gene Symbol: LOC100912165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773296.1 | XP_008771518.1 | 626 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117318 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P536115 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32860 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117318 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085732 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536115 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536115 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117318 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085732 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085732 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117318 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P536115 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P536115 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702029538 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702028596 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62298 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55715 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85246 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89767 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53372 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80551 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15990 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95251 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101689 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702029538 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702028596 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10938469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938478 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938473 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10938467 | - | - |
Rat Genome 230 2.0 Array | 1387199_a_at | - | - |
Rat Genome 230 2.0 Array | 1397853_s_at | - | - |
Rat Genome 230 2.0 Array | 1387199_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1397853_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1397853_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1387199_a_at | - | - |
Affymetrix HT Rat Focus Array | 1397853_s_at | - | - |
Affymetrix HT Rat Focus Array | 1387199_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00011921.rn.1 | TC0X00001882.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011916.rn.1 | TC0X00001882.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011922.rn.1 | TC0X00001882.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00011923.rn.1 | TC0X00001882.rn.1 | RTA TC |
Rat Expression Set 230 | 1387199_PM_a_at | - | - |
Rat Expression Set 230 | 1387199_a_at | - | - |
Rat Expression Set 230 | 1397853_PM_s_at | - | - |
Rat Expression Set 230 | 1397853_s_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938467 | - | - |
Rat Genome U34 Set | rc_AI639196_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22294491 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349180 | - | - |
RatRef-12 Expression BeadChip | ILMN_63975 | - | - |
