Assay Details
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023957.1 | NP_076447.1 | 3 - 4 | 785 | 67 | |
GenBank mRNA | AJ250425.1 | - | 3 - 4 | 785 | 67 | |
AJ302676.1 | - | 2 - 3 | 605 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
