Assay Details
Gene Symbol: Il1rapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177935.2 | NP_808796.1 | 7 - 8 | 1070 | 65 | |
XM_008773261.1 | XP_008771483.1 | 2 - 3 | 830 | 65 | ||
GenBank mRNA | AY216593.1 | - | 7 - 8 | 1070 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
