Assay Details
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106074.1 | NP_001099544.1 | 1 - 2 | 245 | 101 | |
XM_006252872.2 | XP_006252934.1 | 1 - 2 | 288 | 101 | ||
XM_006252874.2 | XP_006252936.1 | 1 - 2 | 288 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
