Assay Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053310.1 | NP_445762.1 | 3 - 4 | 361 | 73 | |
XM_006252890.2 | XP_006252952.1 | 4 - 5 | 487 | 73 | ||
XM_006252891.2 | XP_006252953.1 | 4 - 5 | 484 | 73 | ||
XM_008771094.1 | XP_008769316.1 | 4 - 5 | 485 | 73 | ||
XM_008771095.1 | XP_008769317.1 | 4 - 5 | 485 | 73 | ||
GenBank mRNA | AB020879.2 | - | 3 - 4 | 361 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
