Assay Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053310.1 | NP_445762.1 | 8 - 9 | 948 | 142 | |
XM_006252891.2 | XP_006252953.1 | 9 - 10 | 1071 | 142 | ||
GenBank mRNA | AB020879.2 | - | 8 - 9 | 948 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
