Assay Details
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169100.1 | NP_001162571.1 | 4 - 5 | 429 | 132 | |
XM_006252855.2 | XP_006252917.1 | 4 - 5 | 1349 | 135 | ||
GenBank mRNA | BC158812.1 | - | 4 - 5 | 405 | 132 | |
FQ217580.1 | - | 4 - 5 | 430 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
