Assay Details
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169100.1 | NP_001162571.1 | 5 - 6 | 442 | 131 | |
XM_006252855.2 | XP_006252917.1 | 5 - 6 | 1362 | 134 | ||
GenBank mRNA | BC158812.1 | - | 5 - 6 | 418 | 131 | |
FQ217580.1 | - | 5 - 6 | 443 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
