Assay Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012678.2 | NP_036810.1 | 4 - 5 | 479 | 63 | |
GenBank mRNA | FQ225054.1 | - | 490 | 63 | ||
FQ225082.1 | - | 490 | 63 | |||
J02780.1 | - | 4 - 5 | 482 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
