Assay Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012678.2 | NP_036810.1 | 7 - 8 | 693 | 92 | |
GenBank mRNA | FQ225054.1 | - | 704 | 92 | ||
FQ225082.1 | - | 704 | 92 | |||
J02780.1 | - | 7 - 8 | 696 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
