Assay Details
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201988.2 | NP_973717.1 | 4 - 5 | 452 | 72 | |
GenBank mRNA | AB098134.1 | - | 5 - 6 | 447 | 72 | |
BC098645.1 | - | 4 - 5 | 452 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
