Assay Details
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013880.2 | NP_001013902.2 | 7 - 8 | 1081 | 73 | |
GenBank mRNA | BC079011.1 | - | 5 - 6 | 1085 | 73 | |
CO388828.1 | - | 2 - 3 | 331 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
