Assay Details
Gene Symbol: Tm6sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127654.1 | NP_001121126.1 | 2 - 3 | 208 | 94 | |
GenBank mRNA | BC158807.1 | - | 2 - 3 | 208 | 7324580 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
