Assay Details
Gene Symbol: Cilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107307.1 | NP_001100777.1 | 7 - 8 | 1217 | 152 | |
XM_006252902.2 | XP_006252964.1 | 7 - 8 | 1226 | 152 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
