Assay Details
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252788.2 | XP_006252850.1 | 2 - 3 | 1688 | 130 | |
XM_006252789.2 | XP_006252851.1 | 3 - 4 | 1797 | 130 | ||
XM_008771174.1 | XP_008769396.1 | 2 - 3 | 1796 | 130 | ||
XM_008771175.1 | XP_008769397.1 | 2 - 3 | 1796 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
