Assay Details
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252789.2 | XP_006252851.1 | 7 - 8 | 2444 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
