Assay Details
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012568.2 | NP_036700.1 | 3 - 4 | 818 | 90 | |
XM_003752039.3 | XP_003752087.1 | 4 - 5 | 442 | 90 | ||
XM_008773153.1 | XP_008771375.1 | 3 - 4 | 224 | 90 | ||
GenBank mRNA | AJ310837.1 | - | 3 - 4 | 267 | 90 | |
X61159.1 | - | 3 - 4 | 818 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
