Assay Details
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773209.1 | XP_008771431.1 | 12 - 13 | 1502 | 88 | |
XM_008773210.1 | XP_008771432.1 | 12 - 13 | 1499 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
