Assay Details
Gene Symbol: Cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106603.1 | NP_001100073.1 | 11 - 12 | 2059 | 72 | |
GenBank mRNA | FQ214319.1 | - | 11 - 12 | 1429 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
