Assay Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012096.1 | NP_001012096.1 | 6 - 7 | 619 | 137 | |
XM_006236828.2 | XP_006236890.1 | 7 - 8 | 729 | 137 | ||
XM_006236829.2 | XP_006236891.1 | 7 - 8 | 728 | 137 | ||
XM_006236830.2 | XP_006236892.1 | 6 - 7 | 696 | 137 | ||
XM_006236831.2 | XP_006236893.1 | 7 - 8 | 526 | 137 | ||
GenBank mRNA | BC087064.1 | - | 6 - 7 | 619 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
