Assay Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012096.1 | NP_001012096.1 | 4 - 5 | 461 | 89 | |
XM_006236830.2 | XP_006236892.1 | 4 - 5 | 538 | 89 | ||
GenBank mRNA | BC087064.1 | - | 4 - 5 | 461 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
