Assay Details
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107865.1 | NP_001101335.1 | 10 - 11 | 1256 | 90 | |
XM_006236673.2 | XP_006236735.1 | 11 - 12 | 1565 | 90 | ||
XM_006236674.2 | XP_006236736.1 | 11 - 12 | 1562 | 90 | ||
XM_006236675.2 | XP_006236737.1 | 10 - 11 | 1523 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
