Assay Details
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106610.2 | NP_001100080.2 | 8 - 9 | 723 | 102 | |
GenBank mRNA | BC166430.1 | - | 8 - 9 | 723 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
