Assay Details
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025422.1 | NP_001020593.1 | 10 - 11 | 1304 | 105 | |
XM_008763194.1 | XP_008761416.1 | 10 - 11 | 1348 | 105 | ||
GenBank mRNA | BC098060.1 | - | 10 - 11 | 1304 | 7458113 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
