Assay Details
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025422.1 | NP_001020593.1 | 1 - 2 | 127 | 71 | |
GenBank mRNA | BC098060.1 | - | 1 - 2 | 127 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
