Assay Details
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025422.1 | NP_001020593.1 | 9 - 10 | 1247 | 65 | |
XM_008763194.1 | XP_008761416.1 | 9 - 10 | 1291 | 65 | ||
GenBank mRNA | BC098060.1 | - | 9 - 10 | 1247 | 7458113 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
