Assay Details
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025734.1 | NP_001020905.1 | 8 - 9 | 1417 | 75 | |
XM_006237038.1 | XP_006237100.1 | 9 - 10 | 1884 | 75 | ||
XM_006237040.1 | XP_006237102.1 | 9 - 10 | 1710 | 75 | ||
XM_006237042.1 | XP_006237104.1 | 8 - 9 | 1656 | 75 | ||
GenBank mRNA | BC097414.1 | - | 8 - 9 | 1417 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
