Assay Details
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106602.1 | NP_001100072.1 | 1 - 2 | 196 | 80 | |
XM_006236735.2 | XP_006236797.1 | 1 - 2 | 225 | 80 | ||
XM_006236736.2 | XP_006236798.1 | 1 - 2 | 229 | 80 | ||
XM_006236737.2 | XP_006236799.1 | 1 - 2 | 222 | 80 | ||
XM_008763056.1 | XP_008761278.1 | 1 - 2 | 229 | 80 | ||
XM_008763057.1 | XP_008761279.1 | 1 - 2 | 222 | 80 | ||
GenBank mRNA | BC168743.1 | - | 1 - 2 | 137 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168743.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
