Assay Details
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106602.1 | NP_001100072.1 | 3 - 4 | 500 | 80 | |
XM_006236737.2 | XP_006236799.1 | 3 - 4 | 526 | 80 | ||
XM_008763057.1 | XP_008761279.1 | 3 - 4 | 526 | 80 | ||
GenBank mRNA | BC168743.1 | - | 3 - 4 | 441 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
