Assay Details
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108639.1 | NP_001102109.1 | 8 - 9 | 1015 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P879970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173304 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P879970 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P879970 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P879970 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P452668 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P879970 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57105 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74041 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81638 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10864542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864549 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864545 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864547 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864544 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864548 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864543 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864546 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10864540 | - | - |
Rat Genome 230 2.0 Array | 1374588_at | - | - |
Rat Genome 230 2.0 Array | 1374588_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374588_at | - | - |
Affymetrix HT Rat Focus Array | 1374588_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400024578.rn.1 | TC0400003492.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024577.rn.1 | TC0400003492.rn.1 | RTA TC |
Rat Expression Set 230 | 1374588_PM_at | - | - |
Rat Expression Set 230 | 1374588_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864540 | - | - |
Rat Genome U34 Set | rc_AI030035_at | - | - |
Rat Genome U34 Set | rc_AI169298_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21738993 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360059 | - | - |
