Assay Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236728.2 | XP_006236790.1 | 1 - 2 | 103 | 76 | |
GenBank mRNA | BC087689.1 | - | 1 - 2 | 48 | 7310913 | 76 |
FQ220056.1 | - | 1 - 2 | 47 | 76 | ||
FQ220823.1 | - | 1 - 2 | 48 | 76 | ||
FQ221094.1 | - | 1 - 2 | 47 | 76 | ||
FQ221892.1 | - | 1 - 2 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC087689.1, XM_006236728.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
