Assay Details
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762981.1 | XP_008761203.1 | 6 - 7 | 832 | 103 | |
XM_008762982.1 | XP_008761204.1 | 6 - 7 | 832 | 103 | ||
XM_008762983.1 | XP_008761205.1 | 6 - 7 | 798 | 103 | ||
XM_008762984.1 | XP_008761206.1 | 6 - 7 | 795 | 103 | ||
XM_008762985.1 | XP_008761207.1 | 6 - 7 | 798 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
