Assay Details
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077673.1 | NP_001071141.1 | 5 - 6 | 660 | 114 | |
XM_006237036.1 | XP_006237098.1 | 5 - 6 | 689 | 114 | ||
XM_006237037.1 | XP_006237099.1 | 5 - 6 | 689 | 114 | ||
GenBank mRNA | BC127510.1 | - | 5 - 6 | 660 | 114 | |
EF395817.1 | - | 630 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
cellular response to DNA damage stimulus
spermatogenesis
biological_process
response to UV
negative regulation of DNA recombination
Y-form DNA binding
molecular_function
zinc ion binding
polyubiquitin modification-dependent protein binding
ubiquitin protein ligase binding
DNA repair
cellular response to DNA damage stimulus
spermatogenesis
biological_process
response to UV
negative regulation of DNA recombination
Y-form DNA binding
molecular_function
zinc ion binding
polyubiquitin modification-dependent protein binding
ubiquitin protein ligase binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P23316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61603 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23316 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23316 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P23316 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23316 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70707 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70029 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10864587 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864582 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864580 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864575 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864585 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864578 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864579 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864574 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864573 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864577 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864583 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864581 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864584 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10864572 | - | - |
Rat Genome 230 2.0 Array | 1392449_at | - | - |
Rat Genome 230 2.0 Array | 1392762_at | - | - |
Rat Genome 230 2.0 Array | 1392449_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392762_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392449_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392449_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400024668.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024675.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024677.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024665.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024669.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024673.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024672.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024679.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024666.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024670.rn.1 | TC0400003516.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024674.rn.1 | TC0400003516.rn.1 | RTA TC |
Rat Expression Set 230 | 1392449_at | - | - |
Rat Expression Set 230 | 1392449_PM_at | - | - |
Rat Expression Set 230 | 1392762_at | - | - |
Rat Expression Set 230 | 1392762_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864572 | - | - |
Rat Genome U34 Set | rc_AA800878_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350574 | - | - |
